siRNA Details
| Gene Symbol: GTPBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012227.3 | NP_036359.3 | 2 | 486 | ||
| XM_006724868.3 | XP_006724931.1 | 3 | 386 | |||
| GenBank mRNA | AK024630.1 | - | 2 | 425 | ||
| AK223412.1 | - | 2 | 431 | |||
| AK296003.1 | - | 2 | 147 | |||
| AK298260.1 | - | 2 | 233 | |||
| BC014636.1 | - | 2 | 159 | 3528598 | ||
| BG437484.1 | - | 2 | 569 | |||
| BT007360.1 | - | 1 | 115 | |||
| CN356690.1 | - | 2 | 451 | |||
| DN997384.1 | - | 2 | 406 | |||
| Y14391.2 | - | 2 | 444 | |||

GTPBP6
Genomic Map