siRNA Details
| Gene Symbol: Entpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178106.2 | NP_835207.1 | 5 | 555 | ||
| XM_006244097.2 | XP_006244159.1 | 6 | 915 | |||
| GenBank mRNA | AJ437217.1 | - | 5 | 555 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entpd3| Gene Symbol: Entpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178106.2 | NP_835207.1 | 5 | 555 | ||
| XM_006244097.2 | XP_006244159.1 | 6 | 915 | |||
| GenBank mRNA | AJ437217.1 | - | 5 | 555 | ||