siRNA Details
Gene Symbol: Amigo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178144.1 | NP_835280.1 | 1 | 343 | ||
GenBank mRNA | AY237731.1 | - | 1 | 343 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Amigo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178144.1 | NP_835280.1 | 1 | 343 | ||
GenBank mRNA | AY237731.1 | - | 1 | 343 |