siRNA Details
| Gene Symbol: Sytl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178333.1 | NP_848016.1 | 5 | 647 | ||
| XM_008773069.1 | XP_008771291.1 | 7 | 1977 | |||
| XM_008773070.1 | XP_008771292.1 | 7 | 1968 | |||
| GenBank mRNA | AB098162.1 | - | 5 | 647 | ||

Sytl5
Genomic Map