siRNA Details
| Gene Symbol: Trim50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181080.1 | NP_851594.1 | 1 | 83 | ||
| XM_006249128.2 | XP_006249190.1 | 2 | 405 | |||
| XM_006249129.2 | XP_006249191.1 | 2 | 405 | |||
| XM_006249130.2 | XP_006249192.1 | 2 | 405 | |||
| GenBank mRNA | AY081950.1 | - | 1 | 83 | ||
| BC085684.1 | - | 2 | 243 | 7189609 | ||

Trim50
Genomic Map