siRNA Details
| Gene Symbol: Grik3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF027331.1 | - | 2 | 375 | ||
| M83552.1 | - | 2 | 148 | |||
| Z11716.1 | - | 2 | 375 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grik3| Gene Symbol: Grik3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF027331.1 | - | 2 | 375 | ||
| M83552.1 | - | 2 | 148 | |||
| Z11716.1 | - | 2 | 375 | |||