siRNA Details
| Gene Symbol: Ctsm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181378.2 | NP_852043.1 | 4 | 410 | ||
| GenBank mRNA | AF456462.1 | - | 4 | 410 | ||
| Gene Symbol: RGD1564827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771463.1 | XP_008769685.1 | 3 | 355 | ||

Ctsm
Genomic Map