siRNA Details
| Gene Symbol: Sgms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181386.2 | NP_852051.2 | 10 | 2198 | ||
| GenBank mRNA | AY280961.1 | - | 6 | 1781 | ||
| BC078690.1 | - | 6 | 1543 | 7113038 | ||
| BC081693.1 | - | 10 | 2198 | 7132545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgms1| Gene Symbol: Sgms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181386.2 | NP_852051.2 | 10 | 2198 | ||
| GenBank mRNA | AY280961.1 | - | 6 | 1781 | ||
| BC078690.1 | - | 6 | 1543 | 7113038 | ||
| BC081693.1 | - | 10 | 2198 | 7132545 | ||