siRNA Details
| Gene Symbol: Tsg101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181628.2 | NP_853659.2 | 4 | 319 | ||
| XM_008759341.1 | XP_008757563.1 | 4 | 379 | |||
| GenBank mRNA | AY293306.1 | - | 4 | 278 | ||
| BC070951.1 | - | 4 | 319 | 7100763 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_021884, U52945, AK216304, AK014049, AK179695, and others.

Tsg101
Genomic Map