siRNA Details
Gene Symbol: Gpr135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181771.1 | NP_861436.1 | 1 | 1109 | ||
GenBank mRNA | AY288430.1 | - | 1 | 1109 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
