siRNA Details
Gene Symbol: Tnfaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182950.4 | NP_891995.1 | 6 | 826 | ||
GenBank mRNA | AY336949.2 | - | 6 | 713 | ||
BC128704.1 | - | 6 | 826 | 7459404 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tnfaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182950.4 | NP_891995.1 | 6 | 826 | ||
GenBank mRNA | AY336949.2 | - | 6 | 713 | ||
BC128704.1 | - | 6 | 826 | 7459404 |