siRNA Details
| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183331.1 | NP_899160.1 | 26 | 7697 | ||
| GenBank mRNA | AY362193.1 | - | 26 | 7697 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F8| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183331.1 | NP_899160.1 | 26 | 7697 | ||
| GenBank mRNA | AY362193.1 | - | 26 | 7697 | ||