siRNA Details
| Gene Symbol: Ermp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_184050.2 | NP_908939.2 | 3, 4 | 776 | ||
| XM_006231241.2 | XP_006231303.1 | 3, 4 | 820 | |||
| GenBank mRNA | AY364634.2 | - | 3, 4 | 776 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ermp1| Gene Symbol: Ermp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_184050.2 | NP_908939.2 | 3, 4 | 776 | ||
| XM_006231241.2 | XP_006231303.1 | 3, 4 | 820 | |||
| GenBank mRNA | AY364634.2 | - | 3, 4 | 776 | ||