siRNA Details
| Gene Symbol: Myd88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198130.1 | NP_937763.1 | 3 | 618 | ||
| XM_006244087.2 | XP_006244149.1 | 4 | 965 | |||
| GenBank mRNA | AY191270.1 | - | 3 | 618 | ||
| BC097266.1 | - | 3 | 558 | 7459961 | ||

Myd88
Genomic Map