siRNA Details
| Gene Symbol: Tlr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198131.1 | NP_937764.1 | 2 | 485 | ||
| GenBank mRNA | AY191271.1 | - | 2 | 485 | ||
| AY859725.1 | - | 1 | 389 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tlr9| Gene Symbol: Tlr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198131.1 | NP_937764.1 | 2 | 485 | ||
| GenBank mRNA | AY191271.1 | - | 2 | 485 | ||
| AY859725.1 | - | 1 | 389 | |||