siRNA Details
| Gene Symbol: Gid8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198743.1 | NP_942038.1 | 3 | 209 | ||
| GenBank mRNA | AY156921.1 | - | 3 | 209 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gid8| Gene Symbol: Gid8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198743.1 | NP_942038.1 | 3 | 209 | ||
| GenBank mRNA | AY156921.1 | - | 3 | 209 | ||