siRNA Details
| Gene Symbol: Tpd52l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198744.2 | NP_942039.1 | 2, 3 | 317 | ||
| XM_006235754.2 | XP_006235816.1 | 2, 3 | 316 | |||
| XM_006235757.2 | XP_006235819.1 | 2, 3 | 336 | |||
| XM_006235759.2 | XP_006235821.1 | 2, 3 | 309 | |||
| XM_006235760.2 | XP_006235822.1 | 2, 3 | 331 | |||
| XM_006235761.2 | XP_006235823.1 | 2, 3 | 314 | |||
| XM_008762499.1 | XP_008760721.1 | 2, 3 | 250 | |||
| XM_008762500.1 | XP_008760722.1 | 2, 3 | 248 | |||
| GenBank mRNA | BC059167.1 | - | 2, 3 | 318 | 6890828 | |

Tpd52l2
Genomic Map