siRNA Details
| Gene Symbol: Srr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198757.2 | NP_942052.1 | 3 | 416 | ||
| XM_008768010.1 | XP_008766232.1 | 2 | 331 | |||
| XM_008768011.1 | XP_008766233.1 | 2 | 329 | |||
| XM_008768012.1 | XP_008766234.1 | 2 | 212 | |||
| XM_008768013.1 | XP_008766235.1 | 2 | 212 | |||
| XM_008768014.1 | XP_008766236.1 | 2 | 166 | |||
| XM_008768015.1 | XP_008766237.1 | 2 | 234 | |||
| GenBank mRNA | AB106282.1 | - | 2 | 186 | ||
| BC082014.1 | - | 3 | 416 | 7132082 | ||

Srr
Genomic Map