siRNA Details
| Gene Symbol: Ccdc127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198766.1 | NP_942061.1 | 2 | 178 | ||
| XM_006227783.2 | XP_006227845.1 | 2 | 193 | |||
| GenBank mRNA | BC058157.1 | - | 2 | 178 | 6890640 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc127| Gene Symbol: Ccdc127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198766.1 | NP_942061.1 | 2 | 178 | ||
| XM_006227783.2 | XP_006227845.1 | 2 | 193 | |||
| GenBank mRNA | BC058157.1 | - | 2 | 178 | 6890640 | |