siRNA Details
| Gene Symbol: Zmynd19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198770.2 | NP_942065.1 | 5 | 598 | ||
| XM_006233597.2 | XP_006233659.1 | 596 | ||||
| GenBank mRNA | AJ488144.1 | - | 429 | |||
| BC088093.1 | - | 5 | 598 | 7368043 | ||
| Gene Symbol: LOC100910854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253280.2 | XP_006253342.1 | 1 | 559 | ||

Zmynd19
Genomic Map