siRNA Details
| Gene Symbol: Ppm1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198773.1 | NP_942068.1 | 1, 2 | 572 | ||
| GenBank mRNA | AB081729.1 | - | 1, 2 | 572 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppm1e| Gene Symbol: Ppm1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198773.1 | NP_942068.1 | 1, 2 | 572 | ||
| GenBank mRNA | AB081729.1 | - | 1, 2 | 572 | ||