siRNA Details
Gene Symbol: Tmem110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198774.2 | NP_942069.1 | 7 | 770 | ||
GenBank mRNA | AY283801.1 | - | 7 | 793 | ||
BC088092.1 | - | 7 | 770 | 7368117 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198774.2 | NP_942069.1 | 7 | 770 | ||
GenBank mRNA | AY283801.1 | - | 7 | 793 | ||
BC088092.1 | - | 7 | 770 | 7368117 |