siRNA Details
| Gene Symbol: Tmem138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198777.2 | NP_942072.2 | 5 | 658 | ||
| XM_006231046.1 | XP_006231108.1 | 5 | 751 | |||
| XM_006231047.1 | XP_006231109.1 | 5 | 785 | |||
| XM_006231048.2 | XP_006231110.1 | 5 | 540 | |||
| GenBank mRNA | BC061578.1 | - | 5 | 588 | ||
| BC161808.1 | - | 5 | 930 | 9035805 | ||
| FQ220672.1 | - | 5 | 663 | |||

Tmem138
Genomic Map