siRNA Details
| Gene Symbol: Tmem138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198777.2 | NP_942072.2 | 5 | 683 | ||
| XM_006231046.1 | XP_006231108.1 | 5 | 776 | |||
| XM_006231047.1 | XP_006231109.1 | 5 | 810 | |||
| XM_006231048.2 | XP_006231110.1 | 5 | 565 | |||
| GenBank mRNA | BC061578.1 | - | 5 | 613 | ||
| BC161808.1 | - | 5 | 955 | 9035805 | ||
| FQ220672.1 | - | 5 | 688 | |||

Tmem138
Genomic Map