siRNA Details
| Gene Symbol: Mal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198786.2 | NP_942081.2 | 4 | 692 | ||
| GenBank mRNA | AY079080.1 | - | 4 | 552 | ||
| AY079081.2 | - | 3 | 805 | |||
| BC062388.1 | - | 4 | 692 | 5620891 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mal2| Gene Symbol: Mal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198786.2 | NP_942081.2 | 4 | 692 | ||
| GenBank mRNA | AY079080.1 | - | 4 | 552 | ||
| AY079081.2 | - | 3 | 805 | |||
| BC062388.1 | - | 4 | 692 | 5620891 | ||