siRNA Details
| Gene Symbol: Sdhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198788.2 | NP_942083.1 | 4 | 1052 | ||
| GenBank mRNA | BC059160.1 | - | 4 | 1057 | 6887444 | |
| FQ209438.1 | - | 4 | 1053 | |||
| FQ212791.1 | - | 4 | 1055 | |||
| FQ212873.1 | - | 4 | 1055 | |||
| FQ212985.1 | - | 4 | 1054 | |||
| FQ213509.1 | - | 4 | 1053 | |||
| FQ214315.1 | - | 4 | 1085 | |||
| FQ215255.1 | - | 4 | 1059 | |||
| FQ215355.1 | - | 4 | 1053 | |||
| FQ215505.1 | - | 4 | 1056 | |||
| FQ215951.1 | - | 4 | 1054 | |||
| FQ215986.1 | - | 4 | 1053 | |||
| FQ216027.1 | - | 4 | 1055 | |||
| FQ216314.1 | - | 4 | 1060 | |||
| FQ216503.1 | - | 4 | 1058 | |||
| FQ220862.1 | - | 4 | 1053 | |||
| FQ230166.1 | - | 4 | 1046 | |||

Sdhd
Genomic Map