siRNA Details
| Gene Symbol: Pno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199083.2 | NP_954514.2 | 2, 3 | 398 | ||
| GenBank mRNA | AY344057.1 | - | 2, 3 | 398 | ||
| BC088091.1 | - | 2, 3 | 375 | 7325613 | ||
| FQ215167.1 | - | 2, 3 | 365 | |||
| FQ232331.1 | - | 2, 3 | 378 | |||

Pno1
Genomic Map