siRNA Details
| Gene Symbol: Rcbtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199084.1 | NP_954515.1 | 7 | 643 | ||
| XM_008770845.1 | XP_008769067.1 | 7 | 782 | |||
| XM_008770846.1 | XP_008769068.1 | 7 | 864 | |||
| XM_008770847.1 | XP_008769069.1 | 7 | 827 | |||
| XM_008770848.1 | XP_008769070.1 | 6 | 556 | |||
| XM_008770849.1 | XP_008769071.1 | 6 | 699 | |||
| XM_008770850.1 | XP_008769072.1 | 6 | 781 | |||
| XM_008770851.1 | XP_008769073.1 | 8 | 753 | |||
| XM_008770852.1 | XP_008769074.1 | 8 | 1750 | |||
| XM_008770853.1 | XP_008769075.1 | 8 | 967 | |||
| XM_008770854.1 | XP_008769076.1 | 8 | 735 | |||
| XM_008770855.1 | XP_008769077.1 | 9 | 1131 | |||
| XM_008770856.1 | XP_008769078.1 | 6 | 717 | |||
| XM_008770857.1 | XP_008769079.1 | 5 | 668 | |||
| XM_008770858.1 | XP_008769080.1 | 5 | 623 | |||
| XM_008770859.1 | XP_008769081.1 | 4 | 461 | |||
| XM_008770860.1 | XP_008769082.1 | 4 | 394 | |||
| XM_008770861.1 | XP_008769083.1 | 5 | 536 | |||
| GenBank mRNA | BC061766.1 | - | 7 | 643 | 5621190 | |
| FQ211316.1 | - | 5 | 501 | |||
| FQ211373.1 | - | 5 | 501 | |||

Rcbtb2
Genomic Map