siRNA Details
| Gene Symbol: Spint2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199087.1 | NP_954518.1 | 1, 2 | 398 | ||
| GenBank mRNA | BC061768.1 | - | 1, 2 | 398 | 5621353 | |
| FQ229514.1 | - | 2, 3 | 297 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spint2| Gene Symbol: Spint2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199087.1 | NP_954518.1 | 1, 2 | 398 | ||
| GenBank mRNA | BC061768.1 | - | 1, 2 | 398 | 5621353 | |
| FQ229514.1 | - | 2, 3 | 297 | |||