siRNA Details
| Gene Symbol: Nudt22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199090.2 | NP_954521.1 | 5 | 820 | ||
| XM_006230760.2 | XP_006230822.1 | 4 | 722 | |||
| GenBank mRNA | BC060593.1 | - | 5 | 821 | 6887766 | |
| FQ212766.1 | - | 5 | 767 | |||
| FQ215165.1 | - | 5 | 748 | |||
| FQ219356.1 | - | 5 | 771 | |||

Nudt22
Genomic Map