siRNA Details
Gene Symbol: Svs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199095.1 | NP_954526.1 | 1 | 1534 | ||
XM_006236459.1 | XP_006236521.1 | 2 | 1276 | |||
GenBank mRNA | D00921.1 | - | 1 | 767 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Svs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199095.1 | NP_954526.1 | 1 | 1534 | ||
XM_006236459.1 | XP_006236521.1 | 2 | 1276 | |||
GenBank mRNA | D00921.1 | - | 1 | 767 |