siRNA Details
| Gene Symbol: Zfyve27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199104.2 | NP_954535.2 | 4, 5 | 646 | ||
| XM_006231392.2 | XP_006231454.1 | 4, 5 | 662 | |||
| XM_008760422.1 | XP_008758644.1 | 3, 4 | 1170 | |||
| XM_008760423.1 | XP_008758645.1 | 3, 4 | 1170 | |||
| GenBank mRNA | BC061741.1 | - | 4, 5 | 612 | 5599023 | |

Zfyve27
Genomic Map