siRNA Details
| Gene Symbol: Cnppd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199112.1 | NP_954543.1 | 9 | 1485 | ||
| XM_006245226.2 | XP_006245288.1 | 9 | 1519 | |||
| GenBank mRNA | BC061750.1 | - | 9 | 1485 | 5599704 | |
| FQ232326.1 | - | 1 | 18 | |||

Cnppd1
Genomic Map