siRNA Details
| Gene Symbol: Popdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199113.1 | NP_954544.1 | 4 | 1998 | ||
| GenBank mRNA | AY490241.1 | - | 4 | 1968 | ||
| BC061876.1 | - | 4 | 1998 | 5599130 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Popdc2| Gene Symbol: Popdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199113.1 | NP_954544.1 | 4 | 1998 | ||
| GenBank mRNA | AY490241.1 | - | 4 | 1968 | ||
| BC061876.1 | - | 4 | 1998 | 5599130 | ||