siRNA Details
Gene Symbol: Fgfrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199114.1 | NP_954545.1 | 6 | 1827 | ||
GenBank mRNA | AJ536020.1 | - | 6 | 1827 | ||
BC097261.1 | - | 6 | 1783 | 7442454 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fgfrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199114.1 | NP_954545.1 | 6 | 1827 | ||
GenBank mRNA | AJ536020.1 | - | 6 | 1827 | ||
BC097261.1 | - | 6 | 1783 | 7442454 |