siRNA Details
| Gene Symbol: Ddx24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199119.1 | NP_954550.1 | 2 | 149 | ||
| XM_006240483.1 | XP_006240545.1 | 2 | 352 | |||
| XM_006240484.2 | XP_006240546.1 | 2 | 584 | |||
| GenBank mRNA | AY341876.1 | - | 2 | 149 | ||
| BC097262.1 | - | 2 | 136 | 7442976 | ||

Ddx24
Genomic Map