siRNA Details
| Gene Symbol: Fam21c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199207.1 | NP_954677.1 | 3 | 310 | ||
| XM_006237147.2 | XP_006237209.1 | 3 | 334 | |||
| XM_006237149.2 | XP_006237211.1 | 3 | 334 | |||
| GenBank mRNA | AY257251.1 | - | 3 | 310 | ||

Fam21c
Genomic Map