siRNA Details
| Gene Symbol: Sil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199376.1 | NP_955408.1 | 6 | 608 | ||
| XM_006254542.2 | XP_006254604.1 | 6 | 642 | |||
| XM_008772024.1 | XP_008770246.1 | 6 | 930 | |||
| XM_008772025.1 | XP_008770247.1 | 6 | 694 | |||
| GenBank mRNA | BC062050.1 | - | 6 | 608 | 5598825 | |

Sil1
Genomic Map