siRNA Details
| Gene Symbol: Bpgm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199382.1 | NP_955414.1 | 3 | 1187 | ||
| XM_006236259.2 | XP_006236321.1 | 3 | 1300 | |||
| GenBank mRNA | BC062240.1 | - | 3 | 1187 | 6887677 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bpgm| Gene Symbol: Bpgm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199382.1 | NP_955414.1 | 3 | 1187 | ||
| XM_006236259.2 | XP_006236321.1 | 3 | 1300 | |||
| GenBank mRNA | BC062240.1 | - | 3 | 1187 | 6887677 | |