siRNA Details
| Gene Symbol: Dld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199385.2 | NP_955417.1 | 7 | 600 | ||
| XM_008764590.1 | XP_008762812.1 | 7 | 617 | |||
| GenBank mRNA | BC062069.1 | - | 7 | 604 | 5599215 | |
| FM093110.1 | - | 2 | 178 | |||
| FQ215299.1 | - | 7 | 607 | |||

Dld
Genomic Map