siRNA Details
| Gene Symbol: Snap47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199389.1 | NP_955421.1 | 2 | 571 | ||
| XM_006246488.2 | XP_006246550.1 | 2 | 605 | |||
| GenBank mRNA | BC062056.1 | - | 2 | 571 | 5598911 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snap47| Gene Symbol: Snap47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199389.1 | NP_955421.1 | 2 | 571 | ||
| XM_006246488.2 | XP_006246550.1 | 2 | 605 | |||
| GenBank mRNA | BC062056.1 | - | 2 | 571 | 5598911 | |