siRNA Details
| Gene Symbol: Ica1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199400.1 | NP_955432.1 | 1 | 128 | ||
| XM_006244994.2 | XP_006245056.1 | 2 | 275 | |||
| XM_006244995.2 | XP_006245057.1 | 2 | 275 | |||
| XM_006244998.2 | XP_006245060.1 | 2 | 275 | |||
| XM_006244999.2 | XP_006245061.1 | 2 | 275 | |||
| XM_008767111.1 | XP_008765333.1 | 2 | 275 | |||
| XM_008767112.1 | XP_008765334.1 | 2 | 275 | |||
| GenBank mRNA | AY491990.1 | - | 1 | 128 | ||

Ica1l
Genomic Map