siRNA Details
| Gene Symbol: Suco | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199403.1 | NP_955435.1 | 5, 6 | 593 | ||
| XM_006250164.2 | XP_006250226.1 | 5, 6 | 1048 | |||
| XM_006250166.2 | XP_006250228.1 | 5, 6 | 1048 | |||
| XM_008769657.1 | XP_008767879.1 | 5, 6 | 1048 | |||
| XM_008769658.1 | XP_008767880.1 | 5, 6 | 1048 | |||
| XM_008769659.1 | XP_008767881.1 | 5, 6 | 1048 | |||
| GenBank mRNA | AJ421447.1 | - | 5, 6 | 593 | ||

Suco
Genomic Map