siRNA Details
| Gene Symbol: Man2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199404.1 | NP_955436.1 | 3 | 312 | ||
| XM_006255273.2 | XP_006255335.1 | 3 | 390 | |||
| XM_006255274.2 | XP_006255336.1 | 3 | 389 | |||
| XM_006255275.2 | XP_006255337.1 | 3 | 356 | |||
| GenBank mRNA | BC061819.1 | - | 3 | 312 | 5599365 | |

Man2b1
Genomic Map