siRNA Details
| Gene Symbol: Man2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199404.1 | NP_955436.1 | 11 | 1394 | ||
| GenBank mRNA | BC061819.1 | - | 11 | 1394 | 5599365 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Man2b1| Gene Symbol: Man2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199404.1 | NP_955436.1 | 11 | 1394 | ||
| GenBank mRNA | BC061819.1 | - | 11 | 1394 | 5599365 | |