siRNA Details
| Gene Symbol: Unc5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199407.1 | NP_955439.1 | 2 | 313 | ||
| XM_006233402.2 | XP_006233464.1 | 2 | 261 | |||
| GenBank mRNA | AB118026.1 | - | 2 | 313 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc5c| Gene Symbol: Unc5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199407.1 | NP_955439.1 | 2 | 313 | ||
| XM_006233402.2 | XP_006233464.1 | 2 | 261 | |||
| GenBank mRNA | AB118026.1 | - | 2 | 313 | ||