siRNA Details
| Gene Symbol: Wls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085353.1 | NP_001078822.1 | 2 | 331 | ||
| NM_199408.2 | NP_955440.1 | 2 | 337 | |||
| GenBank mRNA | AY569010.1 | - | 2 | 337 | ||
| BC062391.1 | - | 2 | 331 | 5621215 | ||

Wls
Genomic Map