siRNA Details
| Gene Symbol: Micu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199412.1 | NP_955444.1 | 2 | 133 | ||
| XM_006256420.2 | XP_006256482.1 | 2 | 230 | |||
| XM_006256421.2 | XP_006256483.1 | 2 | 229 | |||
| XM_006256422.2 | XP_006256484.1 | 2 | 229 | |||
| GenBank mRNA | BC062075.1 | - | 2 | 133 | 5599430 | |

Micu1
Genomic Map