siRNA Details
| Gene Symbol: Tgfbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012775.2 | NP_036907.2 | 683 | |||
| XM_006238030.2 | XP_006238092.2 | 697 | ||||
| GenBank mRNA | BC087035.1 | - | 625 | 7098473 | ||
| L26110.1 | - | 652 | ||||
| Gene Symbol: LOC103690035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238153.2 | XP_006238215.2 | 3 | 1968 | ||
| XM_008763706.1 | XP_008761928.1 | 3 | 1980 | |||

Tgfbr1
Genomic Map