siRNA Details
| Gene Symbol: Aqp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142366.2 | NP_001135838.1 | 3 | 571 | ||
| NM_012825.4 | NP_036957.1 | 3 | 563 | |||
| GenBank mRNA | L27588.1 | - | 2 | 431 | ||
| U14007.1 | - | 3 | 497 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aqp4| Gene Symbol: Aqp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142366.2 | NP_001135838.1 | 3 | 571 | ||
| NM_012825.4 | NP_036957.1 | 3 | 563 | |||
| GenBank mRNA | L27588.1 | - | 2 | 431 | ||
| U14007.1 | - | 3 | 497 | |||